Anti-ABL1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12190
10674-174EA 335 USD
10674-174
Anti-ABL1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ABL1/ABL2.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: ABL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR