Anti-ATG4A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22992
10690-384EA 527.69 USD
10690-384
Anti-ATG4A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant ATG4A.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500

Type: Primary
Antigen: ATG4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR