Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-15]

Supplier: Abnova
MAB3831
10705-922EA 406.15 USD
10705-922
Anti-CD14 Mouse Monoclonal Antibody [clone: MEM-15]
Antibodies
Mouse monoclonal antibody raised against full length native CD14.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MEM-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR