Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2]

Supplier: Abnova
MAB3836
10707-726EA 406.15 USD
10707-726
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2]
Antibodies
Mouse monoclonal antibody raised against native CD38.

Recommended Dilutions: Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 ?g/ml; Western Blot: 2 ?g/ml

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR