Anti-CD8A Mouse Monoclonal Antibody [clone: MEM-31]

Supplier: Abnova
MAB3844
10706-842EA 406.15 USD
10706-842
Anti-CD8A Mouse Monoclonal Antibody [clone: MEM-31]
Antibodies
Mouse monoclonal antibody raised against native CD8.

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: MEM-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR