Anti-CACTIN Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23952
10703-830EA 527.69 USD
10703-830
Anti-CACTIN Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C19orf29.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Western Blot: 1:250-1:500

Type: Primary
Antigen: CACTIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR