Anti-GOLM1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16846
10716-110EA 527.69 USD
10716-110
Anti-GOLM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of GOLM1.

Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: GOLM1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR