Anti-Infectious bursal disease virus Mouse Monoclonal Antibody [clone: IBDV67]

Supplier: Abnova
MAB8520
10627-744EA 515 USD
10627-744
Anti-Infectious bursal disease virus Mouse Monoclonal Antibody [clone: IBDV67]
Antibodies
Mouse monoclonal antibody raised against Infectious bursal disease virus (IBDV).

Recommended Dilutions: Recommended pairs for IBD virus in sandwich immunoassay (capture-detection) : IBDV67-IBDV92.

Type: Primary
Antigen: Infectious bursal disease virus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: IBDV67
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR