Anti-Infectious bursal disease virus Mouse Monoclonal Antibody [clone: IBDV99]

Supplier: Abnova
MAB8522
10627-750EA 515 USD
10627-750
Anti-Infectious bursal disease virus Mouse Monoclonal Antibody [clone: IBDV99]
Antibodies
Mouse monoclonal antibody raised against Infectious bursal disease virus (IBDV).

Recommended Dilutions: Recommended pairs for IBD virus in sandwich immunoassay (capture-detection) : IBDV99-IBDV105.

Type: Primary
Antigen: Infectious bursal disease virus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: IBDV99
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR