Anti-SQSTM1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16850
10739-436EA 527.69 USD
10739-436
Anti-SQSTM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of SQSTM1.

  • Recommended Dilutions: Western Blot: 1-2 μg/ml

Type: Primary
Antigen: SQSTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR