Anti-FRMPD3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16853
10715-252EA 527.69 USD
10715-252
Anti-FRMPD3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FRMPD3.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: FRMPD3
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR