Anti-Helicobacter pylori CagA Mouse Monoclonal Antibody [clone: HP-1811]

Supplier: Abnova
MAB8529
10627-598EA 426.15 USD
10627-598
Anti-Helicobacter pylori CagA Mouse Monoclonal Antibody [clone: HP-1811]
Antibodies
Mouse monoclonal antibody raised against partial recombinant Helicobacter pylori CagA.

Type: Primary
Antigen: Helicobacter pylori CagA
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: HP-1811
Epitope:
Host: IgG
Isotype: Mouse
Reactivity: Helicobacter pylori
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR