Anti-Influenza A virus Haemagglutinin H7 Mouse Monoclonal Antibody [clone: 9F2]

Supplier: Abnova
MAB8533
10627-838EA 920 USD
10627-838
Anti-Influenza A virus Haemagglutinin H7 Mouse Monoclonal Antibody [clone: 9F2]
Antibodies
Mouse monoclonal antibody raised against Influenza A haemagglutinin H7 (H7N1).

Type: Primary
Antigen: Influenza A virus Haemagglutinin H7
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 9F2
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR