Anti-COL1A2 Goat Polyclonal Antibody

Supplier: Abnova
PAB14345
10676-000EA 527.69 USD
10676-000
Anti-COL1A2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against native COL1A2.

Recommended Dilutions: Immunocytochemistry: 1:10-1:20; Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: COL1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR