Anti-COL3A1 Goat Polyclonal Antibody

Supplier: Abnova
PAB14346
10676-004EA 527.69 USD
10676-004
Anti-COL3A1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against native COL3A1.

Recommended Dilutions: Immunocytochemistry: 1:10-1:20; Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: COL3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR