Anti-Influenza B virus Mouse Monoclonal Antibody [clone: 15-12]

Supplier: Abnova
MAB8538
10628-224EA 426.15 USD
10628-224
Anti-Influenza B virus Mouse Monoclonal Antibody [clone: 15-12]
Antibodies
Mouse monoclonal antibody raised against influenza B virus.

Type: Primary
Antigen: Influenza B virus
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 15-12
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR