Anti-COL6 Goat Polyclonal Antibody

Supplier: Abnova
PAB14349
10676-010EA 786.15 USD
10676-010
Anti-COL6 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against native COL6.

Recommended Dilutions: Immunocytochemistry: 1:10-1:20; Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: COL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR