Anti-CML Polyclonal Antibody (Biotin)

Supplier: Abnova
PAB14353
10734-828EA 920 USD
10734-828
Anti-CML Polyclonal Antibody (Biotin)
Antibodies
Goat polyclonal antibody raised against CML.

Recommended Dilutions: ELISA: 1:100-1:4000; Western Blot: 1:100-1:4000

Type:
Antigen: CML
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR