Anti-HNE Polyclonal Antibody (Biotin)

Supplier: Abnova
PAB14354
10676-466EA 920 USD
10676-466
Anti-HNE Polyclonal Antibody (Biotin)
Antibodies
Goat polyclonal antibody raised against HNE.

Recommended Dilutions: ELISA: 1:100-1:20000; Western Blot: 1:100-1:20000

Type:
Antigen: HNE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR