Anti-FUT4 Mouse Monoclonal Antibody [clone: 28]

Supplier: Abnova
MAB5622
10715-318EA 406.15 USD
10715-318
Anti-FUT4 Mouse Monoclonal Antibody [clone: 28]
Antibodies
Mouse monoclonal antibody raised against native FUT4.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: FUT4
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 28
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR