Anti-CML Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Abnova
PAB14357
10734-832EA 1046.15 USD
10734-832
Anti-CML Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat polyclonal antibody raised against CML.

Recommended Dilutions: ELISA: 1:100-1:2000; Western Blot: 1:100-1:2000

Type:
Antigen: CML
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR