Anti-TFRC Mouse Monoclonal Antibody [clone: RVS10]

Supplier: Abnova
MAB5627
10741-040EA 406.15 USD
10741-040
Anti-TFRC Mouse Monoclonal Antibody [clone: RVS10]
Antibodies
Mouse monoclonal antibody raised against native TFRC.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: RVS10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR