Anti-APOC3 Goat Polyclonal Antibody (Biotin)

Supplier: Abnova
PAB14367
10680-888EA 653.85 USD
10680-888
Anti-APOC3 Goat Polyclonal Antibody (Biotin)
Antibodies
Goat polyclonal antibody raised against native APOC3.

Recommended Dilutions: ELISA: 1:100-1:10000; Western Blot: 1:100-1:10000

Type:
Antigen: APOC3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR