Anti-GSK3A Monoclonal Antibody [clone: SBGSK3a2]

Supplier: Abnova
MAB5633
10716-634EA 653.85 USD
10716-634
Anti-GSK3A Monoclonal Antibody [clone: SBGSK3a2]
Antibodies
Mouse monoclonal antibody raised against recombinant GSK3A.

Type: Primary
Antigen: GSK3A
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: SBGSK3a2
Epitope:
Host: IgG2a
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR