Anti-MYL2 Mouse Monoclonal Antibody [clone: AT3B2]

Supplier: Abnova
MAB5641
10732-898EA 406.15 USD
10732-898
Anti-MYL2 Mouse Monoclonal Antibody [clone: AT3B2]
Antibodies
Mouse monoclonal antibody raised against full length recombinant MYL2.

Type: Primary
Antigen: MYL2
Clonality: Monoclonal
Clone: AT3B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR