Anti-CD14 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB14381
10705-946EA 406.15 USD
10705-946
Anti-CD14 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CD14.

Recommended Dilutions: Flow Cytometry: 1:100-1:1000

Type: Primary
Antigen: CD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR