Anti-CD14 Mouse Monoclonal Antibody [clone: biG 10]

Supplier: Abnova
MAB5653
10705-900EA 406.15 USD
10705-900
Anti-CD14 Mouse Monoclonal Antibody [clone: biG 10]
Antibodies
Mouse monoclonal antibody raised against CD14.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: biG 10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Horse, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR