Anti-CD14 Mouse Monoclonal Antibody [clone: biG 7]

Supplier: Abnova
MAB5658
10705-914EA 406.15 USD
10705-914
Anti-CD14 Mouse Monoclonal Antibody [clone: biG 7]
Antibodies
Mouse monoclonal antibody raised against CD14.

Recommended Dilutions: ELISA: 1:10000

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: biG 7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR