Anti-PSMB10 Goat Polyclonal Antibody

Supplier: Abnova
PAB14399
10730-406EA 527.69 USD
10730-406
Anti-PSMB10 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of PSMB10.

Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.3-1 ?g/ml.)

Type: Primary
Antigen: PSMB10
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR