Anti-Collagen Type VI Goat Polyclonal Antibody (Biotin)

Supplier: Abnova
PAB14401
10681-108EA 920 USD
10681-108
Anti-Collagen Type VI Goat Polyclonal Antibody (Biotin)
Antibodies
Goat polyclonal antibody raised against native COL6.

Recommended Dilutions: Immunocytochemistry: 1:20-1:40; Immunohistochemistry: 1:20-1:40; ELISA: 1:1000-1:4000

Type:
Antigen: Collagen Type VI
Clonality: Biotin
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR