Anti-LBP Mouse Monoclonal Antibody [clone: biG 33]

Supplier: Abnova
MAB5664
10723-336EA 406.15 USD
10723-336
Anti-LBP Mouse Monoclonal Antibody [clone: biG 33]
Antibodies
Mouse monoclonal antibody raised against recombinant Lbp.

Recommended Dilutions: ELISA: 1:50000

Type: Primary
Antigen: LBP
Clonality: Monoclonal
Clone: biG 33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR