Anti-PHLDA3 Rat Monoclonal Antibody [clone: 4B6]

Supplier: Abnova
MAB6755
10729-114EA 527.69 USD
10729-114
Anti-PHLDA3 Rat Monoclonal Antibody [clone: 4B6]
Antibodies
Rat monoclonal antibody raised against synthetic peptide of PHLDA3.

Type: Primary
Antigen: PHLDA3
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR