Anti-NUP153 Rat Monoclonal Antibody [clone: R4C8]

Supplier: Abnova
MAB6759
10676-948EA 527.69 USD
10676-948
Anti-NUP153 Rat Monoclonal Antibody [clone: R4C8]
Antibodies
Rat monoclonal antibody raised against partial recombinant Nup153.

Type: Primary
Antigen: NUP153
Clonality: Monoclonal
Clone: R4C8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR