Anti-CLCN4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22540
10708-090EA 527.69 USD
10708-090
Anti-CLCN4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CLCN4.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: CLCN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR