Anti-PODXL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

PAB15294
10729-616EA 776.92 USD
10729-616
Anti-PODXL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Rabbit polyclonal antibody raised against synthetic peptide of PODXL.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type:
Antigen: PODXL
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR