Anti-GLO1 Rat Monoclonal Antibody [clone: 6F10]

Supplier: Abnova
MAB6768
10715-966EA 527.69 USD
10715-966
Anti-GLO1 Rat Monoclonal Antibody [clone: 6F10]
Antibodies
Rat monoclonal antibody raised against full length recombinant Glo1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Glo1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 6F10
Epitope:
Host: IgG2b kappa
Isotype: Rat
Reactivity: Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR