Anti-NS3 Mouse Monoclonal Antibody [clone: 20-8]

Supplier: Abnova
MAB6774
10735-066EA 653.85 USD
10735-066
Anti-NS3 Mouse Monoclonal Antibody [clone: 20-8]
Antibodies
Mouse monoclonal antibody raised against partial recombinant Hepatitis C virus NS-3.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: NS3
Clonality: Monoclonal
Clone: 20-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR