Anti-TIMM23 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22549
10741-298EA 527.69 USD
10741-298
Anti-TIMM23 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant TIMM23.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: TIMM23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR