Anti-FAM83B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22556
10714-242EA 527.69 USD
10714-242
Anti-FAM83B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant FAM83B.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: FAM83B
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR