Anti-PDPK1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Abnova
PAB15337
10733-436EA 786.15 USD
10733-436
Anti-PDPK1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PDK1.

Recommended Dilutions: Western Blot: 0.5-2.0 ?g/ml

Type:
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR