Anti-PDHA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Abnova
PAB15338
10687-130EA 653.85 USD
10687-130
Anti-PDHA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PDHA1.

Recommended Dilutions: Western Blot: 1:2500-1:5000

Type:
Antigen: PDHA1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR