Anti-PDHA1 Rabbit Polyclonal Antibody (DyLight® 488)

Supplier: Abnova
PAB15341
10687-124EA 653.85 USD
10687-124
Anti-PDHA1 Rabbit Polyclonal Antibody (DyLight® 488)
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PDHA1.

Recommended Dilutions: Western Blot: 1:2500-1:5000

Type:
Antigen: PDHA1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR