Anti-RP9 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22562
10734-334EA 527.69 USD
10734-334
Anti-RP9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant RP9.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: RP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR