Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 549)

Supplier: Abnova
PAB15345
10707-430EA 786.15 USD
10707-430
Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 549)
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Cdkn2a.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunocytochemistry: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1 ?g/ml

Type:
Antigen: CDKN2A
Clonality: Polyclonal
Clone: 0
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR