Anti-CD19 Mouse Monoclonal Antibody [clone: BU12]

Supplier: Abnova
MAB6901
10705-998EA 406.15 USD
10705-998
Anti-CD19 Mouse Monoclonal Antibody [clone: BU12]
Antibodies
Mouse monoclonal antibody raised against native CD19.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: BU12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR