Anti-CD27 Mouse Monoclonal Antibody [clone: M-T271]

Supplier: Abnova
MAB6903
10706-306EA 406.15 USD
10706-306
Anti-CD27 Mouse Monoclonal Antibody [clone: M-T271]
Antibodies
Mouse monoclonal antibody raised against native CD27.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: M-T271
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR