Anti-CD80 Mouse Monoclonal Antibody [clone: P1.H5.A1.A1]

Supplier: Abnova
MAB6905
10706-860EA 426.15 USD
10706-860
Anti-CD80 Mouse Monoclonal Antibody [clone: P1.H5.A1.A1]
Antibodies
Mouse monoclonal antibody raised against CD80.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: P1.H5.A1.A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR