Anti-PVRL2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB16193
10730-844EA 406.15 USD
10730-844
Anti-PVRL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant PVRL2.

Recommended Dilutions: ELISA: 0.5-1 ?g/ml

Type: Primary
Antigen: PVRL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR