Anti-CD36 Mouse Monoclonal Antibody [clone: SMO]

Supplier: Abnova
MAB6908
10706-406EA 406.15 USD
10706-406
Anti-CD36 Mouse Monoclonal Antibody [clone: SMO]
Antibodies
Mouse monoclonal antibody raised against native CD36.

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: SMO
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR