Anti-MUC17 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22573
10726-232EA 527.69 USD
10726-232
Anti-MUC17 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant MUC17.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: MUC17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR